Merck
  • Anamorelin for patients with cancer cachexia: an integrated analysis of two phase 2, randomised, placebo-controlled, double-blind trials.
Anamorelin for patients with cancer cachexia: an integrated analysis of two phase 2, randomised, placebo-controlled, double-blind trials.
The Lancet. Oncology (2014-12-20)
José M Garcia, Ralph V Boccia, Charles D Graham, Ying Yan, Elizabeth Manning Duus, Suzan Allen, John Friend
ABSTRAKT
Cancer anorexia-cachexia syndrome is associated with increased morbidity and mortality. Anamorelin is an oral ghrelin-receptor agonist with appetite-enhancing and anabolic activity. We assessed the effects of anamorelin on body composition, strength, quality of life, biochemical markers, and safety in patients with cancer anorexia-cachexia. Data were pooled, a priori, from two completed phase 2, multicentre, placebo-controlled, double-blind trials in patients with advanced or incurable cancer and weight loss of 5% or more. Patients were stratified by weight loss severity (5-15%, >15%) and randomly allocated (1:1) with a computer-generated randomisation schedule to anamorelin hydrochloride 50 mg or placebo once-daily for 12 weeks. Primary outcome was lean body mass by dual-energy x-ray absorptiometry over the 12 week treatment period in eligible patients who had at least one dose of study drug and post-treatment efficacy assessment. We assessed safety in all patients who received at least one dose of study drug. The trials are registered with ClinicalTrials.gov, numbers NCT00219817 and NCT00267358. Between June 29, 2005, and Oct 26, 2006, we enrolled 44 patients in the anamorelin group and 38 patients in the placebo group. 74 patients were eligible for the efficacy analyses. Over 12 weeks, lean body mass increased in 38 patients in the anamorelin group by a least-squares mean of 1.89 kg (95% CI 0.84 to 2.95) compared with a decrease of a least-squares mean of -0.20 kg (-1.23 to 0.83) for 36 patients in the placebo group (difference 2.09 kg [0.94-3.25]; p=0.0006). 42 (95%) of 44 patients treated with anamorelin and 33 (87%) of 38 patients treated with placebo had adverse events. The most common grade 3-4 adverse events (treatment-related or not) in the anamorelin group were fatigue, asthenia, atrial fibrillation, and dyspnoea (two [5%] each); in the placebo group, such events were pneumonia (three [8%]) and anaemia, thrombocytopenia, abdominal pain, anxiety, and dyspnoea (two [5%] each). Anamorelin treatment for 12 weeks had a favourable clinical response profile in patients with cancer anorexia-cachexia syndrome. These findings support further investigation in this setting. Helsinn Therapeutics (US), Helsinn Healthcare SA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether