Merck
Cell senescence in myxoid/round cell liposarcoma.
Sarcoma (2014-08-06)
Christina Kåbjörn Gustafsson, Anders Ståhlberg, Katarina Engtröm, Anna Danielsson, Ingela Turesson, Pierre Aman
ABSTRAKT
Myxoid/round cell liposarcoma (MLS/RCLS) is the second most common liposarcoma type and characterized by the fusion oncogenes FUS-DDIT3 or EWSR1-DDIT3. Previous analysis of cell cycle regulatory proteins revealed a prominent expression of G1-cyclins, cyclin dependent kinases, and their inhibitors but very few cells progressing through the G1/S boundary. Here, we extend the investigation to proteins involved in cell senescence in an immunohistochemistry based study of 17 MLS/RCLS cases. Large subpopulations of tumor cells expressed the RBL2 pocket protein and senescence associated heterochromatin 1γ and IL8 receptor β. We conclude that MLS/RCLS tissues contain major populations of senescent tumor cells and this may explain the slow growth rate of this tumor type.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Przeciwciało anty-HP1γ, klon 42s2, clone 42s2, Upstate®, from mouse
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from mouse, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free
Sigma-Aldrich
Anty-p21WAF1 (Ab-1) mysie mAb (EA10), liquid, clone EA10, Calbiochem®