Merck
Chitosan oligosaccharides attenuate ocular inflammation in rats with experimental autoimmune anterior uveitis.
Mediators of inflammation (2014-08-26)
I-Mo Fang, Chang-Hao Yang, Chung-May Yang
ABSTRAKT
We investigated the protective effects and mechanisms of chitosan oligosaccharides (COS) on experimental autoimmune anterior uveitis (EAAU) in rats. EAAU was induced in Lewis rats by footpad and intraperitoneal injections of melanin-associated antigen. The rats received intraperitoneal injections of low-dose (5 mg/kg) or high-dose (10 mg/kg) COS or PBS daily after the immunization. The effects of COS were evaluated by determining the clinical scores and the morphology of the iris/ciliary body (ICB). The expression of inflammatory mediators was evaluated using western blot, immunofluorescence, and ELISA. Treatment with COS significantly attenuated the clinical scores and the leukocyte infiltration in the ICB in a dose-dependent manner. COS effectively reduced the expression of inflammatory mediators (TNF-α, iNOS, MCP-1, RANTES, fractalkine, and ICAM-1). Moreover, COS decreased the IκB degradation and p65 presence in the ICB, which resulted in the inhibition of NF-κB/DNA binding activity. In an in vitro study, sensitized spleen-derived lymphocytes of the COS-treated group showed less chemotaxis toward their aqueous humor and decreased secretion of the above inflammatory mediators in the culture media. COS treated EAAU by inhibiting the activation of NF-κB and reducing the expression of inflammatory mediators. COS might be a potential treatment for acute anterior uveitis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Mouse MCP-1 / CCL2 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human Procalcitonin ELISA Kit
Sigma-Aldrich
Mouse MCP-1 / CCL2 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat MCP-1 / CCL2 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat MCP-1 / CCL2 ELISA Kit, for cell and tissue lysates