Merck
Proteomic analysis of Class IV lupus nephritis.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2014-06-11)
Ayodele Alaiya, Lina Assad, Dania Alkhafaji, Zakia Shinwari, Hadeel Almana, Mohamed Shoukri, Lutfi Alkorbi, Hossamaldin Galal Ibrahim, Mohamed Said Abdelsalam, Edward Skolnik, Chaker Adra, Mamdouh Albaqumi
ABSTRAKT
There have been several attempts to standardize the definition and increase reproducibility in classifying lupus nephritis (LN). The last was made by the International Society of Nephrology and Renal Pathology Society in 2003 where the introduction of Class IV subcategories (global and segmental) was introduced. We investigated whether this subdivision is important using a proteomics approach. All patients with renal biopsies along with their clinical outcome of LN were identified and regrouped according to the above 2003 classifications. Fresh-frozen renal biopsies of Class IV LN (global and segmental), antineutrophil cytoplasmic antibody-associated vasculitis and normal tissue were analyzed using two-dimensional gel electrophoresis (2-DE) and mass spectrometry. Differentially expressed proteins were identified and subjected to principal component analysis (PCA), and post hoc analysis for the four sample groups. PCA of 72 differentially expressed spots separated Class IV global and Class IV segmental from both normal and antineutrophil cytoplasmic antibody-associated vasculitis (ANCA). The 28 identified proteins were used in a post hoc analysis, and showed that IV-global and IV-segmental differ in several protein expression when compared with normal and ANCA. To confirm the proteomic results, a total of 78 patients (50 Class IV-Global and 28 Class IV-Segmental) were re-classified according to 2003 classification. There was no difference in therapy between the groups. The renal survival and patient survivals were similar in both groups. There is no strong evidence to support a different outcome between the two subcategories of Class-IV LN and, they should thus be treated the same until further studies indicate otherwise.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%