Merck
A20 expression in dendritic cells protects mice from LPS-induced mortality.
European journal of immunology (2014-12-05)
Nguyen Thi Xuan, Xu Wang, Gopala Nishanth, Ari Waisman, Katrin Borucki, Berend Isermann, Michael Naumann, Martina Deckert, Dirk Schlüter
ABSTRAKT
DCs contribute to immune homeostasis under physiological conditions and regulate the immune activation during infection. The deubiquitinase A20 inhibits the activation of NF-κB-dependent immune reactions, and prevents the hyperactivation of DCs under steady-state conditions. However, the role of DC-specific A20 under pathological conditions is unknown. Here, we demonstrate that upon injection of low-dose LPS, mice with DC-specific A20 deletion (CD11c-Cre A20(fl/fl) ) died within 6 h, whereas A20(fl/fl) controls survived. LPS-induced mortality in CD11c-Cre A20(fl/fl) mice was characterized by increased serum levels of IL-2, IL-10, IL-12, IFN-γ, and TNF. Upon LPS stimulation, the activation of NF-κB and ERK-NFATc3 pathways were enhanced in A20-deficient DCs, resulting in an increased production of IL-2, IL-12, and TNF both in vitro and in vivo. Targeted inhibition of ERK in A20-deficient DCs abolished the increased production of IL-2. A20-deficient DCs failed to induce LPS tolerance, which was independent of T cells and the intestinal flora, since T-cell depletion and decolonization of CD11c-Cre A20(fl/fl) mice could not prevent death of LPS-challenged CD11c-Cre A20(fl/fl) mice. In conclusion, these findings show that DC-specific A20 preserves immune homeostasis in steady-state conditions and is also required for LPS tolerance.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)