Merck
  • Isolation of a stable subpopulation of mobilized dental pulp stem cells (MDPSCs) with high proliferation, migration, and regeneration potential is independent of age.
Isolation of a stable subpopulation of mobilized dental pulp stem cells (MDPSCs) with high proliferation, migration, and regeneration potential is independent of age.
PloS one (2014-05-30)
Hiroshi Horibe, Masashi Murakami, Koichiro Iohara, Yuki Hayashi, Norio Takeuchi, Yoshifumi Takei, Kenichi Kurita, Misako Nakashima
ABSTRAKT
Insights into the understanding of the influence of the age of MSCs on their cellular responses and regenerative potential are critical for stem cell therapy in the clinic. We have isolated dental pulp stem cells (DPSCs) subsets based on their migratory response to granulocyte-colony stimulating factor (G-CSF) (MDPSCs) from young and aged donors. The aged MDPSCs were efficiently enriched in stem cells, expressing high levels of trophic factors with high proliferation, migration and anti-apoptotic effects compared to young MDPSCs. In contrast, significant differences in those properties were detected between aged and young colony-derived DPSCs. Unlike DPSCs, MDPSCs showed a small age-dependent increase in senescence-associated β-galactosidase (SA-β-gal) production and senescence markers including p16, p21, Interleukin (IL)-1β, -6, -8, and Groα in long-term culture. There was no difference between aged and young MDPSCs in telomerase activity. The regenerative potential of aged MDPSCs was similar to that of young MDPSCs in an ischemic hindlimb model and an ectopic tooth root model. These results demonstrated that the stem cell properties and the high regenerative potential of MDPSCs are independent of age, demonstrating an immense utility for clinical applications by autologous cell transplantation in dental pulp regeneration and ischemic diseases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Staurosporine, VETRANAL®, analytical standard
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture