Merck
Neolignan inhibitors of antigen-induced degranulation in RBL-2H3 cells from the needles of Pinus thunbergii.
Fitoterapia (2014-12-03)
Seong Su Hong, Wonsik Jeong, Jin Kyu Kim, Jin Gwan Kwon, Jae Yeon Lee, Eun-Kyung Ahn, Junho Oh, Dong-Wan Seo, Joa Sub Oh
ABSTRAKT
The ethanolic extract of the needles of Pinus thunbergii was found to suppress antigen mediated degranulation of rat basophilic leukemia (RBL-2H3) cells. A new neolignan glycoside, named pinusthunbergiside A (1), as well as six known neolignan glycosides (2-7) were isolated from the ethanolic extract using bioassay-guided fractionation. Their structures were elucidated by a combination of 1D and 2D NMR, HRESI-MS, and circular dichroism (CD) data. Compounds 2-7 were found for the first time in this plant. The inhibitory effects of isolated constituents on the release of β-hexosaminidase from RBL-2H3 cells were examined, and compounds 2, 3, 5, and 6 were found to show the inhibitory activity with IC₅₀ values ranging between 52.3 and 75.3 μM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tetramethylsilane, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Sigma-Aldrich
PIPES, BioXtra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%