Merck
  • Interplay of the H-bond donor-acceptor role of the distal residues in hydroxyl ligand stabilization of Thermobifida fusca truncated hemoglobin.
Interplay of the H-bond donor-acceptor role of the distal residues in hydroxyl ligand stabilization of Thermobifida fusca truncated hemoglobin.
Biochemistry (2014-12-02)
Francesco P Nicoletti, Juan P Bustamante, Enrica Droghetti, Barry D Howes, Maria Fittipaldi, Alessandra Bonamore, Paola Baiocco, Alessandro Feis, Alberto Boffi, Darío A Estrin, Giulietta Smulevich
ABSTRAKT
The unique architecture of the active site of Thermobifida fusca truncated hemoglobin (Tf-trHb) and other globins belonging to the same family has stimulated extensive studies aimed at understanding the interplay between iron-bound ligands and distal amino acids. The behavior of the heme-bound hydroxyl, in particular, has generated much interest in view of the relationships between the spin-state equilibrium of the ferric iron atom and hydrogen-bonding capabilities (as either acceptor or donor) of the OH(-) group itself. The present investigation offers a detailed molecular dynamics and spectroscopic picture of the hydroxyl complexes of the WT protein and a combinatorial set of mutants, in which the distal polar residues, TrpG8, TyrCD1, and TyrB10, have been singly, doubly, or triply replaced by a Phe residue. Each mutant is characterized by a complex interplay of interactions in which the hydroxyl ligand may act both as a H-bond donor or acceptor. The resonance Raman stretching frequencies of the Fe-OH moiety, together with electron paramagnetic resonance spectra and MD simulations on each mutant, have enabled the identification of specific contributions to the unique ligand-inclusive H-bond network typical of this globin family.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O