Merck
Source apportionment of polycyclic aromatic hydrocarbons in PM2.5 using positive matrix factorization modeling in Shanghai, China.
Environmental science. Processes & impacts (2014-12-11)
Fengwen Wang, Tian Lin, Jialiang Feng, Huaiyu Fu, Zhigang Guo
ABSTRAKT
Providing quantitative information on the sources of PM2.5-bound polycyclic aromatic hydrocarbons (PAHs) in urban regions is vital to establish effective abatement strategies for air pollution in a megacity. In this study, based on a year data set from October 2011 to August 2012, the sources of PM2.5-bound 16 USEPA priority PAHs (16 PAHs) in Shanghai, a megacity in China, were apportioned by positive matrix factorization (PMF) modeling. The average concentrations (in ng m(-3)) of 16 PAHs in PM2.5 in the fall, winter, spring and summer were 20.5 ± 18.2, 27.2 ± 24.0, 13.7 ± 7.7 and 6.4 ± 8.1, respectively, with an annual average of 16.9 ± 9.0. The source apportionment by PMF indicated that coal burning (30.5%) and gasoline engine emission (29.0%) were the two major sources of PAHs in the PM2.5 in Shanghai, followed by diesel engine emission (17.5%), air-surface exchange (11.9%) and biomass burning (11.1%). The highest source contributor for PAHs in the fall and winter was gasoline engine emission (36.7%) and coal burning (41.9%), respectively; while in the spring and summer, it was diesel engine emission that contributed the most (52.1% and 43.5%, respectively). It was suggested that there was a higher contribution of PAHs from engine emissions in 2011-2012 compared with those in 2002-2003. The major sources apportioned by PMF complemented well with this of using diagnostic ratios, suggesting a convincing identification of sources for the PM2.5-bound 16 PAHs in a megacity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acenaphthylene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, analytical standard
Supelco
Acenaphthylene, analytical standard
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Acenaphthylene, 75%
Supelco
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acenaphthene, analytical standard
Sigma-Aldrich
Acenaphthene, 99%
Supelco
Standardowa temperatura topnienia 79-81°C, analytical standard
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland