Merck
  • Purification of two triterpenoids from Schisandra chinensis by macroporous resin combined with high-speed counter-current chromatography.
Purification of two triterpenoids from Schisandra chinensis by macroporous resin combined with high-speed counter-current chromatography.
Journal of chromatographic science (2013-11-14)
Lijie Zhu, Bin Li, Xiuying Liu, Xianjun Meng
ABSTRAKT
A method for preparative purification of corosolic acid and nigranoic acid from Schisandra chinensis (SC) was established using a combination of macroporous absorption resin column separation and high-speed counter-current chromatography (HSCCC). The crude extracts obtained from SC using 70% ethanol were separated on a macroporous resin column and then eluted with a graded ethanol series. The 70% ethanol fraction was used as the sample for separation of the two triterpenoids by HSCCC. The two-phase solvent system used for HSCCC separation was chloroform-n-butanol-methanol-water (10:0.5:7:4, v/v/v/v). The upper phase was used as the stationary phase of HSCCC. Corosolic acid (16.4 mg) of 96.3% purity and nigranoic acid (9.5 mg) of 98.9% purity were obtained in a one-step HSCCC separation from 100 mg of the sample. The structures of corosolic acid and nigranoic acid were identified by (1)H-nuclear magnetic resonance (NMR) and (13)C-NMR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material