Merck
Snail1-dependent transcriptional repression of Cezanne2 in hepatocellular carcinoma.
Oncogene (2013-06-25)
Z Xu, L Pei, L Wang, F Zhang, X Hu, Y Gui
ABSTRAKT
High malignancy and early metastasis are the hallmarks of hepatocellular carcinoma (HCC). Here, we report that Cezanne2 expression is downregulated in HCC cells and in HCC patients' tumorous tissues and that Cezanne2 is inversely associated with Snail1 expression in HCC patients' tumorous tissues. Chromatin immunoprecipitation assays and the reporter gene assay showed that Snail1 binds to the promoter of the Cezanne2 gene and mediates the direct consequence of Cezanne2 repression. Enhanced expression of Cezanne2 could suppress proliferation, migration and invasion in HCC cells. Further, Cezanne2 could regulate MMP (matrix metalloproteinase)2, MMP9 and ICAM1 (intercellular adhesion molecule) levels through modulation of the NF-κB (nuclear factor kappa-light-chain-enhancer of activated B cell) signaling cascade. Co-immunoprecipitation and in vivo deubiquitination assay indicated that Cezanne2 interacts with TNF receptor-associated factor (TRAF)6 and cleaves the polyubiquitin from TRAF6 substrates. Our data reveal that Snail1-mediated suppression of Cezanne2 may have a key role in HCC malignancy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine