Merck
  • The secretion of IL-6 by CpG-ODN-treated cancer cells promotes T-cell immune responses partly through the TLR-9/AP-1 pathway in oral squamous cell carcinoma.
The secretion of IL-6 by CpG-ODN-treated cancer cells promotes T-cell immune responses partly through the TLR-9/AP-1 pathway in oral squamous cell carcinoma.
International journal of oncology (2014-03-29)
Min Ruan, Katherine Thorn, Shengwen Liu, Siyi Li, Wenjun Yang, Chunye Zhang, Chenping Zhang
ABSTRAKT
Increasing evidence suggests that communication between tumor and immune cells can alter the tumor microenvironment in ways that promote tumor development. The purpose of this study was to characterize the immune response elicited by TLR-9-activated OSCC cells, to identify the cytokines involved in the signaling pathway and to elucidate the molecular mechanism of this pathway in OSCC cells. MTS, flow cytometry and ELISA assay were used to evaluate T-cell immune responses, cancer cell proliferation and pro-inflammatory cytokine secretion, respectively. Western blot analysis, EMSA and ChIP assay were employed to detect the activity of the NF-κB and AP-1 signaling pathways. A marked response was observed when T-cells were co-cultured with supernatants from CpG-ODN-treated OSCC cells. This response was characterized by increased CD4+ and CD8+ T-cell proliferation and an increase in IFN-γ production by the CD4+ T-cell population. Treatment of OSCC cells with CpG-ODN resulted in an increase in IL-6 secretion as well as an increase in AP-1 binding activity to the IL-6 promoter. Moreover, blockage of the TLR-9/AP-1 pathway significantly decreased IL-6 expression and T-cell immune response. In human OSCC, the TLR-9 pathway, when stimulated by CpG-ODNs, promotes a T-cell immune response mediated by AP-1-activated IL-6 secretion. Although the complete molecular mechanism has yet to be understood, these findings provide evidence linking tumor cell activities to immune system responses. In addition, the TLR-9/AP-1/IL-6 pathway provides new therapeutic targets for the prevention and treatment of OSCC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Curcumin, primary reference standard
USP
Curcumin, United States Pharmacopeia (USP) Reference Standard
Supelco
Curcumin, analytical standard
Supelco
Curcumin, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC)