Merck
  • Simultaneous determination of bosentan and glimepiride in human plasma by ultra performance liquid chromatography tandem mass spectrometry and its application to a pharmacokinetic study.
Simultaneous determination of bosentan and glimepiride in human plasma by ultra performance liquid chromatography tandem mass spectrometry and its application to a pharmacokinetic study.
Journal of pharmaceutical and biomedical analysis (2014-04-02)
Xiangjun Qiu, Jianlong Zhao, Zhe Wang, Zhisheng Xu, Ren-ai Xu
ABSTRAKT
A sensitive and rapid ultra performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS) method was developed to determine bosentan (BOS) and glimepiride (GPD) in human plasma simultaneously. Chromatographic separation was carried out on an Acquity UPLC BEH C18 column and mass spectrometric analysis was performed using a QTrap5500 mass spectrometer coupled with an electro-spray ionization (ESI) source in the positive ion mode. The MRM transitions of m/z 552.0→202.1 and m/z 491.2→125.9 were used to quantify BOS and GPD, respectively. This assay method has been fully validated in terms of selectivity, linearity, recovery and matrix effect, accuracy, precision and stability. The linearity of this method was found to be within the concentration range of 5-1000 ng/mL for BOS, and 2.5-500 ng/mL for GPD in human plasma. Only 1.5 min was needed for an analytical run. This assay was used to support a clinical study where multiple oral doses were administered to healthy Chinese subjects to investigate the pharmacokinetics of BOS and GPD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%