Merck
Loss of sulfur dioxide and changes in some chemical properties of Malatya apricots (Prunus armeniaca L.) during sulfuring and drying.
Journal of the science of food and agriculture (2014-01-22)
Meltem Türkyılmaz, Mehmet Özkan, Nihal Güzel
ABSTRAKT
This study was conducted to determine the differences in some analytical properties of four apricot cultivars and to determine the changes in these analytical properties during sulfuring and sun-drying. There were significant differences in the contents of polyphenols, carotenoids and organic acids (OA) as well as antioxidant activities (AOAs) of the cultivars (P < 0.05). After sulfuring and drying, considerable reductions were detected in the contents of total polyphenols (TPCs, 11-26%), OAs (4-32%) and β-carotene (6-21%), and AOAs (2-21%) of the samples. Sun-drying resulted in 71-83% decreases in sulfur dioxide (SO2 ) contents of sulfured-dried apricots (SDAs) in comparison with apricots immediately after sulfuring. As the TPCs increased, the SO2 absorption by the samples also increased. In contrast, the OA contents had no effect on SO2 absorption, but an increase in OA content resulted in an increase in the browning values of the SDAs. As expected, increases in contents of ferulic acid (r = 0.932), chlorogenic acid (r = 0.850), epicatechin (r = 0.804) and quercetin (r = 0.750) led to an increase in browning values of the SDAs. There were significant effects of cultivar and processing on the physico-chemical properties investigated in the study, and with the absorption of SO2 and the formation of a brown colour in the samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Kwercetyna, United States Pharmacopeia (USP) Reference Standard
Supelco
Malic acid, Pharmaceutical Secondary Standard; Certified Reference Material