Merck
Single vial freeze-dried TRODAT-1 kit: preparation and demonstration of clinical efficacy of [(99m)Tc]TRODAT-1 in Indian scenario.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2014-12-06)
Aruna Korde, Mythili Kameswaran, Drishty Satpati, Haladhar Dev Sarma, Sharmila Banerjee, Ajit Shinto, K K Kamaleshwaran, Grace Samuel
ABSTRAKT
A single vial freeze-dried kit formulation for preparation of three patients' dose of [(99m)Tc]TRODAT-1 has been developed for early diagnosis of Parkinson's disease (PD). Kits were evaluated to ascertain the purity, stability and batch to batch variations. Preclinical evaluation was carried out in laboratory animals and clinical imaging was performed in human patients with PD. The labeling yield and purity of [(99m)Tc]TRODAT-1 was >90%. Swiss mice showed retention of [(99m)Tc]TRODAT-1 in the mid brain region. Clinical studies showed decreased striatal uptake with increasing severity of PD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tin(II) chloride dihydrate, reagent grade, 98%
Sigma-Aldrich
Tin(II) chloride dihydrate, ACS reagent, 98%
Supelco
Tin(II) chloride dihydrate, suitable for AAS
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.97% trace metals basis
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Tantalum(V) ethoxide, packaged for use in deposition systems
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%