Merck
OATP and MRP2-mediated hepatic uptake and biliary excretion of eprosartan in rat and human.
Pharmacological reports : PR (2014-06-10)
Pengyuan Sun, Changyuan Wang, Qi Liu, Qiang Meng, Aijie Zhang, Xiaokui Huo, Huijun Sun, Kexin Liu
ABSTRAKT
Eprosartan is an angiotensin II receptor antagonist, used in the treatment of hypertension and heart failure in clinical patients. The objective of this study was to clarify the mechanism underlying hepatic uptake and biliary excretion of eprosartan in rats and humans. Perfused rat liver in situ, rat liver slices, isolated rat hepatocytes and human organic anion-transporting polypeptide (OATP)-transfected cells in vitro were used in this study. Extraction ratio of eprosartan was decreased by rifampicin in perfused rat livers. Uptake of eprosartan in rat liver slices and isolated rat hepatocytes was significantly inhibited by Oatp modulators such as ibuprofen, digoxin, rifampicin and cyclosporine A, but not by tetraethyl ammonium or p-aminohippurate. Uptake of eprosartan in rat hepatocytes indicated a saturable process. Although uptake of eprosartan in OATP1B3-human embryonic kidney cells (HEK) 293 cells was not observed, significant differences in cellular accumulations of eprosartan between vector- and OATP1B1-Madin-Darby canine kidney strain (MDCK) II cells were found in transcellular transport study. Moreover, cumulative biliary excretion rate of eprosartan in the presence of probenecid (Multidrug resistance-associated protein 2 (Mrp2) inhibitor) was significantly decreased in perfused rat livers. Vectorial basal-to-apical transport of eprosartan was also observed in OATP1B1/MRP2 double transfectants. Eprosartan was transported by multiple Oatps (at least Oatp1a1 and Oatp1a4)/Mrp2 in rat and OATP1B1/MRP2, at least, in human.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
USP
Cilostazol, United States Pharmacopeia (USP) Reference Standard
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Cilostazol, ≥98% (HPLC), powder
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Probenecid, European Pharmacopoeia (EP) Reference Standard
USP
Probenecid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Probenecid
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%