Merck
Reaction of acetaldehyde with 5-aminolevulinic acid via dihydropyrazine derivative.
Chemical & pharmaceutical bulletin (2015-03-10)
Toshinori Suzuki, Naoki Yasuhara, Takashi Ueda, Michiyo Inukai, Mitsunobu Mio
ABSTRAKT
When a solution of 5-aminolevulinic acid (ALA) was incubated with acetaldehyde at neutral pH, a product was generated. This product was identified as 3-ethylpyrazine-2,5-dipropanoic acid (ETPY). ETPY was stable at neutral pH. It has been reported that ALA dimerizes at neutral pH generating 3,6-dihydropyrazine-2,5-dipropanoic acid (DHPY), and subsequently resulting in pyrazine-2,5-dipropanoic acid (PY) by autoxidation. In the present reaction, DHPY generated from ALA reacted with acetaldehyde, resulting in ETPY. Preadministration of ALA 3 min prior to acetaldehyde injection supressed the toxicity of acetaldehyde in male mice. These results suggest that ALA may be useful as a scavenger for acetaldehyde.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥97.0% (AT)
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, PESTANAL®, analytical standard