Merck
  • Orthogonal zirconium diol/C18 liquid chromatography-tandem mass spectrometry analysis of poly and perfluoroalkyl substances in landfill leachate.
Orthogonal zirconium diol/C18 liquid chromatography-tandem mass spectrometry analysis of poly and perfluoroalkyl substances in landfill leachate.
Journal of chromatography. A (2014-08-12)
B McKay Allred, Johnsie R Lang, Morton A Barlaz, Jennifer A Field
ABSTRAKT
Leachates coming from landfills contain a myriad of compounds of potential environmental and human health concern, including per- and polyfluorinated alkyl substances (PFASs). Micro liquid-liquid extraction was combined with a 900μl large volume injection (LVI) for the analysis of 70 PFASs in landfill leachate by orthogonal LC-MS/MS. The LVI approach introduced 7 times more extract than conventional injection approaches. Two zirconium-modified diol guard columns effectively retained PFASs from the organic leachate extract and an analytical C18 column was used for separation. Method accuracy and precision for PFASs with analytical grade reference materials ranged from 81-120% and 5.5-33%, respectively. Estimated method detection limits in the low to sub-ng/L. Seven landfill leachates were analyzed by the optimized analytical method for the purposes of method demonstration. Leachates were characterized by a wide variety of PFASs, reporting on 36 previously-unanalyzed PFASs in leachate. Perfluoroalkyl carboxylates were the most abundant class detected while phosphorous-containing PFASs, present in all leachates, were at low concentrations. The 3-perfluoropentyl propanoate (5:3 FTCA) was the most concentrated analyte in most samples and constitutes a previously unreported but significant component of landfill leachate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard