Merck
Prostacyclin-synthase expression in head and neck carcinoma patients and its prognostic value in the response to radiotherapy.
The Journal of pathology (2014-09-27)
Mercedes Camacho, Zenaida Piñeiro, Sonia Alcolea, Jacinto García, Josep Balart, Ximena Terra, Francesc-Xavier Avilés-Jurado, Marta Soler, Miquel Quer, Xavier León, Luis Vila
ABSTRAKT
Prostacyclin (PGI2 ) plays a role in cancer progression but the mechanism is currently poorly understood. Additionally, no data are available about the prognostic value of the PGI2 pathway in head and neck squamous cell carcinoma (HNSCC) therapy. We evaluated the expression of the PGI2 pathway in HNSCC patients. PGI2 production and PGI synthase (PGIS) expression, in terms of mRNA (RT-PCR) and protein (immunoblotting), were lower in tumour samples than in non-tumoural mucosa, whereas, as expected, COX-2 expression was increased in HNSCC tumour samples. Using local control of the tumour after radiotherapy or chemoradiotherapy as a dependent variable, patients were classified into two categories of PGIS transcript levels. The high-PGIS group had a significantly lower frequency of local and distant failure than the low-PGIS group, and the 5-year cancer-specific survival was higher [90.2% (95% CI 81.0-99.4%) versus 60.5% (95% CI 44.4-76.6%)]. None of the four HNSCC cell lines analysed expressed PGIS and therefore they did not produce PGI2 . However, HNSCC-conditioned media enhanced PGI2 production in endothelial cells (ECs). The stable analogue of PGI2 , carbaprostacyclin (cPGI2 ), exerted little effect on HNSCC cell line migration, and no effect on cell cycle distribution or proliferation rate after radiation injury was observed. Nevertheless, cPGI2 promoted EP-4-dependent in vitro angiogenesis. Von Willebrand factor expression (EC marker) and capillary density were significantly higher in the group of patients with high expression of PGIS. Our results indicate that PGIS expression was associated with radiotherapy efficiency. Although we do not provide direct evidence of a relationship between tumour vascularization and radiotherapy efficiency, our results suggest that the effect of PGI2 is related to its ability to promote vascularization. These results also support the concept that co-adjuvant therapy with PGIS enhancers, such as retinoids, could have therapeutic value for HNSCC treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O