Merck
Electron paramagnetic resonance imaging for real-time monitoring of Li-ion batteries.
Nature communications (2015-02-11)
M Sathiya, J-B Leriche, E Salager, D Gourier, J-M Tarascon, H Vezin
ABSTRAKT
Batteries for electrical storage are central to any future alternative energy paradigm. The ability to probe the redox mechanisms occurring at electrodes during their operation is essential to improve battery performances. Here we present the first report on Electron Paramagnetic Resonance operando spectroscopy and in situ imaging of a Li-ion battery using Li2Ru0.75Sn0.25O3, a high-capacity (>270 mAh g(-1)) Li-rich layered oxide, as positive electrode. By monitoring operando the electron paramagnetic resonance signals of Ru(5+) and paramagnetic oxygen species, we unambiguously prove the formation of reversible (O2)(n-) species that contribute to their high capacity. In addition, we visualize by imaging with micrometric resolution the plating/stripping of Li at the negative electrode and highlight the zones of nucleation and growth of Ru(5+)/oxygen species at the positive electrode. This efficient way to locate 'electron'-related phenomena opens a new area in the field of battery characterization that should enable future breakthroughs in battery research.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylene carbonate, 98%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propylene carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Propylene carbonate, suitable for HPLC, 99.7%
Sigma-Aldrich
Dimethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Dimethyl carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Hexafluoropropene, ≥99%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.