Merck
Probing the conformational changes of ovalbumin after glycation using HDX-MS.
Food chemistry (2014-07-24)
Xiaoqin Huang, Zongcai Tu, Hui Wang, Qiuting Zhang, Yuan Chen, Yan Shi, Hui Xiao
ABSTRAKT
The conformational changes of the glycated ovalbumin were studied by hydrogen/deuterium exchange coupled with high resolution mass spectrometry technique (HDX-MS). After incubation with glucose at 50 °C for 6 h, 9 glycated peptides were detected and the corresponding glycation sites were identified. The glycation extent of each peptide was relatively high, almost over 0.5 in all peptides. A detailed peptide mapping revealed that most of the peptides, including the glycated and non-glycated were protected. The glycation sites not only influence the local region but also the distant area. The enhanced hydrogen protection after glycation suggests that the protein adopts a more stable conformation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D