Merck
Multi-electrode array capable of supporting precisely patterned hippocampal neuronal networks.
Biomedical microdevices (2015-02-06)
Tianyi Zhou, Susan F Perry, Yevgeny Berdichevsky, Susanne Petryna, Vicki Fluck, Svetlana Tatic-Lucic
ABSTRAKT
Accurate positioning of primary mouse hippocampal neurons on electrodes enables the recording from and stimulation of specified individual neurons on a multi-electrode array (MEA). In this work, positive dielectrophoresis (DEP) is applied to actively recruit hippocampal neurons to the electrodes of a MEA, whereas microstructures such as chambers and trenches are created to effectively define a patterned neuronal network. We present here the effective pretreatment methods, to improve cytocompatibility of cured thin SU-8 epoxy, commonly used in the fabrication of MEAs. The functionality of our novel MEA is proven by the successful recording of spontaneous and stimulated neuronal potentials from primary hippocampal neurons, including the propagation of evoked neuronal bursts between electrodes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)