Merck
Cooperative therapeutic action of retinoic acid receptor and retinoid x receptor agonists in a mouse model of Alzheimer's disease.
Journal of Alzheimer's disease : JAD (2014-06-12)
Kohichi Kawahara, Michita Suenobu, Hideyuki Ohtsuka, Akihiko Kuniyasu, Yukihiko Sugimoto, Madoka Nakagomi, Hiroshi Fukasawa, Koichi Shudo, Hitoshi Nakayama
ABSTRAKT
Alzheimer's disease (AD) is a neurodegenerative process involving amyloid-β (Aβ) peptide deposition, neuroinflammation, and progressive memory loss. Here, we evaluated whether oral administration of retinoic acid receptor (RAR)α,β agonist Am80 (tamibarotene) or specific retinoid X receptor (RXR) pan agonist HX630 or their combination could improve deficits in an AD model, 8.5-month-old amyloid-β protein precursor 23 (AβPP23) mice. Co-administration of Am80 (0.5 mg/kg) and HX630 (5 mg/kg) for 17 days significantly improved memory deficits (Morris water maze) in AβPP23 mice, whereas administration of either agent alone produced no effect. Only co-administration significantly reduced the level of insoluble Aβ peptide in the brain. These results thus indicate that effective memory improvement via reduction of insoluble Aβ peptide in 8.5-month-old AβPP23 mice requires co-activation of RARα,β and RXRs. RARα-positive microglia accumulated Aβ plaques in the AβPP23 mice. Rat primary microglia co-treated with Am80/HX630 showed increased degradation activity towards 125I-labeled oligomeric Aβ1-42 peptide in an insulin-degrading enzyme (IDE)-dependent manner. The co-administration increased mRNA for IDE and membrane-associated IDE protein in vivo, suggesting that IDE contributes to Aβ clearance in Am80/HX630-treated AβPP23 mice. Am80/HX630 also increased IL-4Rα expression in microglial MG5 cells. The improvement in memory of Am80/HX630-treated AβPP23 mice was correlated with the levels and signaling of hippocampal interleukin-4 (IL-4). Therefore, Am80/HX630 may promote differentiation of IL-4-responsive M2-like microglia and increase their activity for clearance of oligomeric Aβ peptides by restoring impaired IL-4 signaling in AβPP23 mice. Combination treatment with RAR and RXR agonists may be an effective approach for AD therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
GW9662, >98% (HPLC)
Sigma-Aldrich
Anty-IDE, N-końcowe (97-273) królicze pAb, liquid, Calbiochem®