Merck
  • In situ derivatization combined to automated microextraction by packed sorbents for the determination of chlorophenols in soil samples by gas chromatography mass spectrometry.
In situ derivatization combined to automated microextraction by packed sorbents for the determination of chlorophenols in soil samples by gas chromatography mass spectrometry.
Journal of chromatography. A (2014-08-13)
Rosa María González Paredes, Carmelo García Pinto, José Luis Pérez Pavón, Bernardo Moreno Cordero
ABSTRAKT
A method based on the coupling of in situ extraction and derivatization of chlorophenols (CPs) (2-chlorophenol, 4-chloro-3-methylphenol, 2,4-dichlorophenol, and 2,4,6-trichlorophenol) from soils, accomplishing their preconcentration by means of automated microextraction by packed sorbent (MEPS), is proposed. After extraction and acylation of the chlorophenols in aqueous medium, the liquid phase obtained is subjected to the MEPS procedure. The QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) and MEPS techniques were compared and the results confirmed the preconcentration carried out with MEPS. The existence of a matrix effect was checked and the analytical characteristics of the method were determined in a soil sample. The method provided good linearity (from 1 to 12μgkg(-1)), together with good repeatability and reproducibility values (RSD equal to or less than 10%). The limits of detection were in the 0.118-0.894μgkg(-1) range. A certified reference material was applied to validate the proposed methodology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ethyl acetate, analytical standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%