Merck
  • Characterization of the novel positive allosteric modulator of the metabotropic glutamate receptor 4 ADX88178 in rodent models of neuropsychiatric disorders.
Characterization of the novel positive allosteric modulator of the metabotropic glutamate receptor 4 ADX88178 in rodent models of neuropsychiatric disorders.
The Journal of pharmacology and experimental therapeutics (2014-06-21)
Mikhail Kalinichev, Emmanuel Le Poul, Christelle Boléa, Françoise Girard, Brice Campo, Massimiliano Fonsi, Isabelle Royer-Urios, Susan E Browne, Jason M Uslaner, Matthew J Davis, Jacob Raber, Robert Duvoisin, Simon T Bate, Ian J Reynolds, Sonia Poli, Sylvain Celanire
ABSTRAKT
There is growing evidence that activation of metabotropic glutamate receptor 4 (mGlu4) leads to anxiolytic- and antipsychotic-like efficacy in rodent models, yet its relevance to depression-like reactivity remains unclear. Here, we present the pharmacological evaluation of ADX88178 [5-methyl-N-(4-methylpyrimidin-2-yl)-4-(1H-pyrazol-4-yl)thiazol-2-amine], a novel potent, selective, and brain-penetrant positive allosteric modulator of the mGlu4 receptor in rodent models of anxiety, obsessive compulsive disorder (OCD), fear, depression, and psychosis. ADX88178 dose-dependently reduced the number of buried marbles in the marble burying test and increased open-arm exploration in the elevated plus maze (EPM) test, indicative of anxiolytic-like efficacy. Target specificity of the effect in the EPM test was confirmed using male and female mGlu4 receptor knockout mice. In mice, ADX88178 reduced the likelihood of conditioned freezing in the acquisition phase of the fear conditioning test, yet had no carryover effect in the expression phase. Also, ADX88178 dose-dependently reduced duration of immobility in the forced swim test, indicative of antidepressant-like efficacy. ADX88178 reduced DOI (2,5-dimethoxy-4-iodoamphetamine)-mediated head twitches (albeit with no dose-dependency), and MK-801 [(5S,10R)-(+)-5-methyl-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5,10-imine]-induced locomotor hyperactivity in mice, but was inactive in the conditioned avoidance response test in rats. The compound showed good specificity as it had no effect on locomotor activity in mice and rats at efficacious doses. Thus, allosteric activation of mGlu4 receptors can be a promising new therapeutic approach for treatment of anxiety, OCD, fear-related disorders, and psychosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
(±)-Baclofen, ≥98% (HPLC), solid
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Olanzapine, European Pharmacopoeia (EP) Reference Standard
USP
Olanzapine, United States Pharmacopeia (USP) Reference Standard
Diazepam, European Pharmacopoeia (EP) Reference Standard
Olanzapine for system suitability, European Pharmacopoeia (EP) Reference Standard
Diazepam for system suitability, European Pharmacopoeia (EP) Reference Standard
Baclofen, European Pharmacopoeia (EP) Reference Standard