Merck
Western diet in ApoE-LDLR double-deficient mouse model of atherosclerosis leads to hepatic steatosis, fibrosis, and tumorigenesis.
Laboratory investigation; a journal of technical methods and pathology (2014-09-10)
Marian Kampschulte, Christiane Stöckl, Alexander C Langheinrich, Ulrike Althöhn, Rainer M Bohle, Gabriele A Krombach, Philipp Stieger, Yuri Churin, Sandra Kremer, Christian Dierkes, Timo Rath, Elke Roeb, Martin Roderfeld
ABSTRAKT
Nonalcoholic fatty liver disease has been linked to cardiovascular diseases and atherosclerosis. The aim of the current study was to characterize the hepatic pathology leading to fibrosis and tumors in a murine model of atherosclerosis. Male apolipoprotein E/low-density lipoprotein receptor double-knockout mice (AL) mice were fed with a high fat and high cholesterol western diet for 35 weeks (AL mice on WD). Protein and mRNA analysis as well as micro-computed tomography (micro-CT) were performed to assess oxidative stress, liver damage, inflammation, fibrosis, signaling pathways, vascularization, and tumorigenesis. Controls were chosen to distinguish between genetically and dietary effects in steatohepatitis and associated tumorigenesis. Hepatic inflammation and dyslipidemia were increased in AL mice on WD compared with wild-type mice on WD. Uniquely, AL mice on WD showed a spontaneous development of tumors (30% of cases) and thickening of intrahepatic vessel walls. Functionally relevant underlying signaling pathways such as NF-κB, Stat3, JNK, and AKT were differentially regulated between AL and wild-type mice on WD. Micro-CT was capable of visualizing and quantitatively distinguishing tumor neovascularization from vascularization in non-neoplastic liver tissue. AL mice on WD diet represent a novel model combining atherosclerosis and nonalcoholic fatty liver disease. Signaling pathways of liver cell damage and compensatory liver regeneration in combination with enhanced inflammation appear to be crucial for the spontaneous development of tumors in AL mice on WD. Micro-CT represents a new and powerful technique for the ultrastructural and three-dimensional assessment of the vascular architecture of liver tumors.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications