Merck
Analytical and microbiological characterization of paper samples exhibiting foxing stains.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada (2015-03-20)
Margarida Nunes, Cátia Relvas, Francisca Figueira, Joana Campelo, António Candeias, Ana T Caldeira, Teresa Ferreira
ABSTRAKT
This work comprises the use of a multi-analytical approach combined with microbiological studies to characterize six paper samples, containing foxing stains, from the 20th century, regarding their cellulose matrix, fillers, and sizing materials, and to evaluate possible paper degradation that might have occurred during the foxing stains. Photography under different illuminations and optical microscopy were used for morphological characterization of the paper samples and foxing stains. Scanning electron microscopy coupled energy dispersive spectroscopy (SEM-EDS) was of particular importance for defining the presence of fiber disorder and disruption on the surface of some of the stains, and localized accumulations of mineral-like particles on the surface of others. SEM-EDS, attenuated total reflection Fourier transform infrared spectroscopy (ATR-FT-IR), and energy dispersive X-ray fluorescence (EDXRF) were used for the identification of mineral fillers, whereas sizing agents were analyzed using ATR-FT-IR. EDXRF results showed that no differences, within the standard deviation, were found in iron and copper contents between the foxed and unfoxed areas. Fungi belonging to the genus Penicillium spp. were found in all the paper samples. Unfoxed areas presented lower contamination than the foxed areas.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology