Merck
Stability-indicating RP-HPLC method for the quantitative analysis of perindopril erbumine in tablet dosage form.
Journal of chromatographic science (2013-05-22)
Hari Hara Theja Dugga, Ramalingam Peraman, Devanna Nayakanti
ABSTRAKT
A specific, stability-indicating reversed-phase high-performance liquid chromatography (RP-HPLC) method was developed and validated for the estimation of perindopril erbumine (PDE) in tablet dosage form. The HPLC method showed adequate separation of PDE from its degradation products. The separation was achieved on a Phenomenex Luna C18 column (250 × 4.6 mm × 5 µm) using a mobile phase composition of 0.2% trifluoroacetic acid buffer and acetonitrile in the ratio of 60:40 (pH adjusted to 3 with ammonia) at a flow rate of 1 mL/min. The injection volume was 20 µL and the wavelength of detection was kept at 215 nm. Stress studies were performed with 1 mg/mL of each drug, starting with mild conditions and followed by stronger conditions to achieve sufficient degradation at approximately 5-20%. The linearity of the proposed method was investigated in the range of 2.5 to 50 µg/mL for PDE. The limits of detection and quantification were found to be 0.75 and 2.3 µg/mL, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%