Merck
  • Differential effects of ketoconazole, itraconazole and voriconazole on the pharmacokinetics of imatinib and its main metabolite GCP74588 in rat.
Differential effects of ketoconazole, itraconazole and voriconazole on the pharmacokinetics of imatinib and its main metabolite GCP74588 in rat.
Drug development and industrial pharmacy (2013-09-24)
Gaotong Lin, Chenming Wang, Xiangjun Qiu, Zhe Wang, Anyue Han, Tao Xu, Xin Kan, Guoxin Hu
ABSTRAKT
The aim of the study was to develop a high performance liquid chromatography method for simultaneous determination of imatinib and CGP74588 in rat serum and study the inhibition effects of ketoconazole, itraconazole and voriconazole on pharmacokinetics of imatinib and CGP74588 in rats. In our study, we found that ketoconazole caused a significant increase (63.4%) in the AUC of imainib and a 28.8% increase in Cmax, which was greater than that of itraconazole but lower than that of voriconazole. When co-administered with voriconazole, pharmacokinetic parameters of imatinib were not significantly altered except for a 36.8% increase in the Cmax of imtinib. The Cmax of CGP74588 was decreased by 55.8% and AUC(0-∞) 49.7%, while the Vz/F and CLz/F values were increased by 1.7-fold and 1.1-fold, respectively. Itraconazole did not significantly influence the pharmacokinetic parameters of imatinib and CGP74588. The difference may be related to the different variation of inhibition sites of the three azole antifungal agents on CYP3A4 and P-gp. In clinical, when imatinib was co-administrated with ketoconazole or voriconazole, dose adjustment of imatinib should be taken into account.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%