Merck
Modulation of gene expression by 3-iodothyronamine: genetic evidence for a lipolytic pattern.
PloS one (2014-11-08)
Veronica Mariotti, Erika Melissari, Caterina Iofrida, Marco Righi, Manuela Di Russo, Riccardo Donzelli, Alessandro Saba, Sabina Frascarelli, Grazia Chiellini, Riccardo Zucchi, Silvia Pellegrini
ABSTRAKT
3-Iodothyronamine (T1AM) is an endogenous biogenic amine, structurally related to thyroid hormone, which is regarded as a novel chemical messenger. The molecular mechanisms underlying T1AM effects are not known, but it is possible to envisage changes in gene expression, since delayed and long-lasting phenotypic effects have been reported, particularly with regard to the modulation of lipid metabolism and body weight. To test this hypothesis we analysed gene expression profiles in adipose tissue and liver of eight rats chronically treated with T1AM (10 mg/Kg twice a day for five days) as compared with eight untreated rats. In vivo T1AM administration produced significant transcriptional effects, since 378 genes were differentially expressed in adipose tissue, and 114 in liver. The reported changes in gene expression are expected to stimulate lipolysis and beta-oxidation, while inhibiting adipogenesis. T1AM also influenced the expression of several genes linked to lipoprotein metabolism suggesting that it may play an important role in the regulation of cholesterol homeostasis. No effect on the expression of genes linked to toxicity was observed. The assay of tissue T1AM showed that in treated animals its endogenous concentration increased by about one order of magnitude, without significant changes in tissue thyroid hormone concentration. Therefore, the effects that we observed might have physiological or pathophysiological importance. Our results provide the basis for the reported effectiveness of T1AM as a lipolytic agent and gain importance in view of a possible clinical use of T1AM in obesity and/or dyslipidaemia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)