Merck
Molecular recognition of HER-1 in whole-blood samples.
Journal of molecular recognition : JMR (2014-10-04)
Iuliana Moldoveanu, Camelia Stanciu Gavan, Raluca-Ioana Stefan-van Staden
ABSTRAKT
Multimode sensing was proposed for molecular screening and recognition of HER-1 in whole blood. The tools used for molecular recognition were platforms based on nanostructured materials such as the complex of Mn(III) with meso-tetra (4-carboxyphenyl) porphyrin, and maltodextrin (dextrose equivalence between 4 and 7), immobilized in diamond paste, graphite paste or C60 fullerene paste. The identification of HER-1 in whole-blood samples, at molecular level, is performed using stochastic mode and is followed by the quantification of it using stochastic and differential pulse voltammetry modes. HER-1 can be identified in the concentration range between 280 fg/ml and 4.86 ng/ml using stochastic mode, this making possible the early detection of cancers such as gastrointestinal, pancreatic and lung cancers. The recovery tests performed using whole-blood samples proved that the platforms can be used for identification and quantification of HER-1 with high sensitivity and reliability in such samples, these making them good molecular screening tools.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%