Merck
Maldi-tof fingerprinting of seminal plasma lipids in the study of human male infertility.
Lipids (2014-06-18)
Mariana Camargo, Paula Intasqui, Camila Bruna de Lima, Daniela Antunes Montani, Marcílio Nichi, Eduardo Jorge Pilau, Fabio Cesar Gozzo, Edson Guimarães Lo Turco, Ricardo Pimenta Bertolla
ABSTRAKT
This study proposed lipid fingerprinting of human seminal plasma by mass spectrometry as an analytical method to differentiate biological conditions. For this purpose, we chose infertile men as a model to study specific conditions, namely: high and low seminal plasma lipid peroxidation levels (sub-study 1.1), high and low sperm nuclear DNA fragmentation (sub-study 1.2), and intervention status: before and after subinguinal microsurgical varicocelectomy (study 2). Study 1 included 133 patients, of which 113 were utilized for sub-study 1.1 and 89 for sub-study 1.2. Study 2 included 17 adult men submitted to subinguinal varicocelectomy, before and 90 days after varicocelectomy. Lipids were extracted from seminal plasma and submitted to Matrix-Assisted Laser Desorption Ionization Quadrupole-Time-of-Flight Mass Spectrometry in the positive ionization mode. Spectra were processed using Waters(®) MassLynx, and MetaboAnalyst online software was used for statistical analyses. For sub-studies 1.1 and 1.2, and study 2, univariate analysis revealed 8, 87 and 34 significant ions, respectively. Multivariate analysis was performed through PCA and PLS-DA. PCA generated 56, 32 and 34 components respectively for each study and these were submitted to logistic regression. A ROC curve was plotted and the area under the curve was equal to 97.4, 92.5 and 96.5%. PLS-DA generated a list of 19, 24 and 23 VIP ions for sub-studies 1.1 and 1.2, and study 2, respectively. Therefore, this study established the lipid profile and comparison of patterns altered in response to specific biological conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Supelco
2,5-Dihydroxybenzoic acid, analytical standard