Merck
  • Fabrication of flexible, transparent and conductive films from single-walled carbon nanotubes with high aspect ratio using poly((furfuryl methacrylate)-co-(2-(dimethylamino)ethyl methacrylate)) as a new polymeric dispersant.
Fabrication of flexible, transparent and conductive films from single-walled carbon nanotubes with high aspect ratio using poly((furfuryl methacrylate)-co-(2-(dimethylamino)ethyl methacrylate)) as a new polymeric dispersant.
Nanoscale (2015-03-26)
Taeheon Lee, Byunghee Kim, Sumin Kim, Jong Hun Han, Heung Bae Jeon, Young Sil Lee, Hyun-Jong Paik
ABSTRAKT
We synthesized poly((furfuryl methacrylate)-co-(2-(dimethylamino)ethyl methacrylate)) (p(FMA-co-DMAEMA)) for the dispersion of single-walled carbon nanotubes (SWCNTs) while maintaining their high aspect ratios. The nanotubes' length and height were 2.0 μm and 2 nm, as determined by transmission electron microscopy and atomic force microscopy, respectively. Transparent conductive films (TCFs) were fabricated by individually dispersed long SWCNTs onto a flexible polyethylene terephthalate substrate. The sheet resistance (Rs) was 210 Ω □(-1) with 81% transmittance at a wavelength of 550 nm. To reduce their Rs, the TCFs were treated with HNO3 and SOCl2. After treatment, the TCFs had an Rs of 85.75 Ω □(-1) at a transmittance of 85%. The TCFs exhibited no appreciable change over 200 repeated bending cycles. Dispersing SWCNTs with this newly synthesized polymer is an effective way to fabricate a transparent, highly conductive and flexible film.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
4,4′-Dinonyl-2,2′-dipyridyl, 97%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
VenPure® SF, powder
Supelco
Furfural, analytical standard
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl α-bromoisobutyrate, 98%
Sigma-Aldrich
Copper(I) bromide, 98%
Sigma-Aldrich
Copper(I) bromide, AnhydroBeads, -10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Copper(I) bromide, 99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
2-(Dimethylamino)ethyl methacrylate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-(Dimethylamino)ethyl methacrylate, contains 700-1000 ppm monomethyl ether hydroquinone as inhibitor, 98%