Merck
Platelet hyperactivity, neurobehavioral symptoms and depression among Indian women chronically exposed to low level of arsenic.
Neurotoxicology (2014-12-03)
Bidisha Mukherjee, Banani Bindhani, Hirak Saha, Dona Sinha, Manas Ranjan Ray
ABSTRAKT
The prevalence of neurobehavioral symptoms (NBS) and depression has been investigated in premenopausal rural women of West Bengal, India enrolled from arsenic (As) endemic (groundwater As 11-50 μg/L; n = 342) and control areas (As level ≤ 10 μg/L; n = 312). The subjective symptoms questionnaire and Beck's 21-point depression inventory-II were used for the detection of NBS and depression, respectively. Platelet P-selectin expression was measured by flow cytometry, plasma neurotransmitter activity with high performance liquid chromatography and groundwater As level by atomic absorption spectroscopy. The As level in groundwater was 2.72 ± 1.18 μg/L in control and 28.3 ± 13.51 μg/L in endemic areas (p < 0.0001). Women residing in endemic areas demonstrated a higher prevalence of depressive symptoms (39.8 vs. 19.9%, p < 0.001) and anxiety (43.3 vs. 18.0% in control, p < 0.001), fatigue (68.4 vs. 23.4%, p < 0.0001), reduced sense of taste (15.8 vs. 4.5%, p<0.0001) and smell (14.9 vs. 5.8%, p < 0.001); burning sensation (36.8 vs. 5.4%, p < 0.0001) and tingling or numbness in the extremities (25.1 vs. 5.1%, p < 0.0001); and transient loss of memory (69.9 vs. 28.2%, p < 0.001). As-exposed women had 1.6-times more plasma epinephrine and norepinephrine (p < 0.05), 1.8-times higher level plasma serotonin with 28.9% lower intraplatelet serotonin (p < 0.05 for both), but their plasma dopamine level was not significantly different (p>0.05) from that of controls. Moreover, women from endemic areas had 2.3-times more P-selectin-expressing platelets in their circulation (p < 0.001). After controlling the potential confounders, chronic low level As (11-50 μg/L) exposure showed a positive association with the prevalence of neurobehavioral symptoms and depression among Indian women in their child-bearing age.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, analytical standard
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard