Merck
Suppression of memory acquisition following co-administration of lithium and atorvastatin through nitric oxide pathway in mice.
Pharmacology, biochemistry, and behavior (2014-04-09)
Amir Reza Honarmand, Nasim Pourtabatabaei, Nastaran Rahimi, Ahmad Reza Dehpour, Mehrak Javadi-Paydar
ABSTRAKT
The aim of this study was to investigate the interactive effect of lithium and atorvastatin on cognitive performance and the role of NO as a potential mechanism involved in this interaction. Memory performance was evaluated in a two-trial recognition Y-maze test and a step-through passive avoidance task in mice. Lithium (5, 10, 20 or 40 mg/kg, i.p.) and atorvastatin (1 mg/kg, p.o.) were administered 1 h before each trial, L-NAME, a non-specific NO synthase inhibitor (3, 10 mg/kg, i.p.); aminoguanidine, a specific inducible NO synthase (iNOS) inhibitor (100 mg/kg); and L-arginine, a NO precursor (750 mg/kg) were administered 30 min before training sessions. The level of plasma NO end-products (NOx) was determined using Griess reagent protocol. 1) Lithium (40 mg/kg) impaired the acquisition of spatial recognition memory; 2) lithium did not affect the retrieval phase of spatial memory; 3) atorvastatin (1 mg/kg) significantly impaired the memory performance, when co-administered with the sub-effective dose of lithium (10 mg/kg), but did not affect the status when administered with lithium (5 mg/kg); 4) L-NAME (10 mg/kg) and aminoguanidine (100 mg/kg) dramatically decreased memory performance in mice received sub-effective doses of both lithium (5 mg/kg) and atorvastatin (1 mg/kg); 5) L-arginine (750 mg/kg) improved the memory acquisition in mice administered lithium (10 mg/kg) and atorvastatin (1 mg/kg); 6) lithium did not affect the cognitive performance in the passive avoidance test. All results were compatible and confirmed with in vitro determination of plasma NOx levels. Lithium, dose dependently, impaired acquisition phase of spatial recognition memory. Lithium and atorvastatin co-administration impaired spatial recognition memory mediating by nitrergic pathway. In addition to L-arginine, our data from L-NAME and aminoguanidine also support the involvement of NO pathway in this interaction.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Supelco
Roztwór elektrolitu, nonaqueous, LiCl in ethanol (saturated)
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Lithium chloride solution, 2 M in ethanol