Merck
  • A dispersive liquid-liquid micellar microextraction for the determination of pharmaceutical compounds in wastewaters using ultra-high-performace liquid chromatography with DAD detection.
A dispersive liquid-liquid micellar microextraction for the determination of pharmaceutical compounds in wastewaters using ultra-high-performace liquid chromatography with DAD detection.
Biomedical chromatography : BMC (2014-07-25)
Sarah Montesdeoca-Esponda, Cristina Mahugo-Santana, Zoraida Sosa-Ferrera, José Juan Santana-Rodríguez
ABSTRAKT
A dispersive liquid-liquid micellar microextraction (DLLMME) method coupled with ultra-high-performance liquid chromatography (UHPLC) using Diode Array Detector (DAD) detector was developed for the analysis of five pharmaceutical compounds of different nature in wastewaters. A micellar solution of a surfactant, polidocanol, as extraction solvent (100 μL) and chloroform as dispersive solvent (200 μL) were used to extract and preconcentrate the target analytes. Samples were heated above critical temperature and the cloudy solution was centrifuged. After removing the chloroform, the reduced volume of surfactant was then injected in the UHPLC system. In order to obtain high extraction efficiency, the parameters affecting the liquid-phase microextraction, such as time and temperature extraction, ionic strength and surfactant and organic solvent volume, were optimized using an experimental design. Under the optimized conditions, this procedure allows enrichment factors of up to 47-fold. The detection limit of the method ranged from 0.1 to 2.0 µg/L for the different pharmaceuticals. Relative standard deviations were <26% for all compounds. The procedure was applied to samples from final effluent collected from wastewater treatment plants in Las Palmas de Gran Canaria (Spain), and two compounds were measured at 67 and 113 µg/L in one of them.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Nonaethylene glycol monododecyl ether, nonionic surfactant
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Ornidazole, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, analytical standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Ketoprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketoprofen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
USP
Standardowa temperatura topnienia kofeiny, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra