Merck
A new prenylated flavanone from Derris trifoliata Lour.
Molecules (Basel, Switzerland) (2012-01-13)
Cheng Jiang, Shengzi Liu, Weihong He, Xiongming Luo, Si Zhang, Zhihui Xiao, Ximin Qiu, Hao Yin
ABSTRAKT
A new flavanone, 4',5,7-trihydroxy-6,8-di-(2-hydroxy-3-methylbut-3-enyl)- flavanone, was isolated from the aerial parts of Derris trifoliate, together with eleven known compounds: rotenone, tephrosin, 12a-hydroxyrotenone, deguelin, 6a,12a-dehydro-rotenone, dehydrodeguelin, 7a-O-methyldeguelol, 7a-O-methylelliptonol, 5,7,3',4'-tetra-hydroxy-6,8-diprenylisoflavone, daidzein and 4'-hydroxy-7-methoxyflavanone. 7a-O-Methylelliptonol was isolated for the first time from the genus Derris. Their structures were characterized on the basis of spectral data. Eight of the isolated compounds were found to be significantly toxic to brine shrimp (LC(50) range 0.06-9.95 μg/mL). The new compound showed weak toxicity (LC(50) = 211.31 μg/mL).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4′,5,7-Trihydroxy-6,8-diprenylisoflavone, ≥85% (LC/MS-UV)