Merck
Automatic determination of NET (neutrophil extracellular traps) coverage in fluorescent microscopy images.
Bioinformatics (Oxford, England) (2015-03-21)
Luis Pedro Coelho, Catarina Pato, Ana Friães, Ariane Neumann, Maren von Köckritz-Blickwede, Mário Ramirez, João André Carriço
ABSTRAKT
Neutrophil extracellular traps (NETs) are believed to be essential in controlling several bacterial pathogens. Quantification of NETs in vitro is an important tool in studies aiming to clarify the biological and chemical factors contributing to NET production, stabilization and degradation. This estimation can be performed on the basis of fluorescent microscopy images using appropriate labelings. In this context, it is desirable to automate the analysis to eliminate both the tedious process of manual annotation and possible operator-specific biases. We propose a framework for the automated determination of NET content, based on visually annotated images which are used to train a supervised machine-learning method. We derive several methods in this framework. The best results are obtained by combining these into a single prediction. The overall Q(2) of the combined method is 93%. By having two experts label part of the image set, we were able to compare the performance of the algorithms to the human interoperator variability. We find that the two operators exhibited a very high correlation on their overall assessment of the NET coverage area in the images (R(2) is 97%), although there were consistent differences in labeling at pixel level (Q(2), which unlike R(2) does not correct for additive and multiplicative biases, was only 89%). Open source software (under the MIT license) is available at https://github.com/luispedro/Coelho2015_NetsDetermination for both reproducibility and application to new data.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder