Merck
Characterization of the interaction between superoxide dismutase and 2-oxoisovalerate dehydrogenase.
Gene (2015-05-11)
Chongfu Song, Hebin Li, Liangquan Sheng, Xiaobo Zhang
ABSTRAKT
Thermophiles are attractive microorganisms to study the adaptation of life in high temperature environment. It is revealed that superoxide dismutase (SOD) is essential for thermoadaptation of thermophiles. However, the SOD-mediated pathway of thermoadaptation remains unclear. To address this issue, the proteins interacted with SOD were characterized in Thermus thermophilus in this study. Based on co-immunoprecipitation and Western blot analyses, the results showed that 2-oxoisovalerate dehydrogenase α subunit was bound to SOD. The isothermal titration calorimetry analysis showed the existence of the interaction between SOD and 2-oxoisovalerate dehydrogenase α subunit. The bacterial two-hybrid data indicated that SOD was directly interacted with 2-oxoisovalerate dehydrogenase α subunit. Gene site-directed mutagenesis analysis revealed that the intracellular interaction between SOD and 2-oxoisovalerate dehydrogenase α subunit was dependent on their whole molecules. Therefore our study presented a novel aspect of SOD in the thermoadaptation of thermophiles by interaction with dehydrogenase, a key enzyme of tricarboxylic acid cycle.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder