Merck
  • UPLC-ESI-Q-TOF-MS(E) and GC-MS identification and quantification of non-intentionally added substances coming from biodegradable food packaging.
UPLC-ESI-Q-TOF-MS(E) and GC-MS identification and quantification of non-intentionally added substances coming from biodegradable food packaging.
Analytical and bioanalytical chemistry (2015-07-04)
Elena Canellas, Paula Vera, Cristina Nerín
ABSTRAKT
Biodegradable packagings are made by combination of several materials creating a multilayer with the properties needed. Each material, including the adhesive, could contain substances that could migrate to the food. In this work, gas chromatography coupled with mass spectrometry and ultra-high-pressure liquid chromatography coupled with quadrupole time-of-flight mass spectrometry were used to identify the biodegradable adhesive compounds. Five of the 13 compounds identified were nonintentionally added substances; they were neoformed compounds created by the reaction of added compounds in the adhesive. Moreover, the migration of the compounds through four different biodegradable materials-paper, polylactic acid, ecovio®, and polyvinyl alcohol-was studied for the first time. Three of the 13 compounds identified in the adhesive migrated from the adhesive to Tenax®, which was used as a solid food simulant. One of them, 2,4,7,9-tetramethyl-5-decyne-4,7-diol, was an intentionally added substance, and the other two were 1,6-dioxacyclododecane-7,12-dione and 1,6,13,18-tetraoxacyclotetracosane-7,12,19,24-tetraone, which were nonintentionally added substances identified in this work. Higher migration values (ranging from 0.81 to 2.07 mg/kg) were observed for migration through ecovio® than through the multilayer made by combination of ecovio® and polyvinyl alcohol (0.07-0.39 mg/kg) owing to the barrier effect provided by polyvinyl alcohol. The migration values for migration through paper and polylactic acid were below the limits of detection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-Benzisothiazol-3(2H)-one, 97%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Giemsa stain, technical grade, used as a blood stain
Sigma-Aldrich
Giemsa stain, certified by the BSC
Sigma-Aldrich
Adipic acid, BioXtra, ≥99.5% (HPLC)
Sigma-Aldrich
Adipic acid, 99%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder