Merck
Generation of a soluble recombinant trimeric form of bovine CD40L and its potential use as a vaccine adjuvant in cows.
Veterinary immunology and immunopathology (2015-11-11)
Julien Pujol, Fabrice Bouillenne, Frédéric Farnir, Isabelle Dufrasne, Jacques Mainil, Moreno Galleni, Pierre Lekeux, Fabrice Bureau, Laurence Fiévez
ABSTRAKT
Vaccination is the most cost-effective way to control infectious diseases in cattle. However, many infectious diseases leading to severe economical losses worldwide still remain for which a really effective and safe vaccine is not available. These diseases are most often due to intracellular pathogens such as bacteria or viruses, which are, by their localization, protected from antibiotics and/or CD4(+) T cell-dependent humoral responses. We therefore postulated that strategies leading to induction of not only CD4(+) T cell responses but also CD8(+) cytotoxic T lymphocyte (CTL) responses against infected cells should be privileged in the development of new vaccines against problematic intracellular pathogens in bovines. CD40 signaling in antigen-presenting cells may lead to the induction of robust CD4-independent CTL responses and several studies, especially in mice, have used CD40 stimulation to promote CD8(+) T cell-mediated immunity. For example, we have recently shown that immunization of mice with heat-killed Staphylococcus aureus (HKSA) and agonistic anti-CD40 monoclonal antibodies elicits strong CTL responses capable of protecting mice from subsequent staphylococcal mastitis. Unfortunately, there is at present no tool available to efficiently stimulate CD40 in cattle. In this study, we therefore first produced a soluble recombinant trimeric form of the natural bovine CD40 ligand (sboCD40LT). We then observed that sboCD40LT was able to potently stimulate bovine cells in vitro. Finally, we provide evidence that immunization of cows with sboCD40LT combined with HKSA was able to significantly increase the number of both HKSA-specific CD4(+) and CD8(+) T cells in the draining lymph nodes. In conclusion, we suggest that this new molecular tool could help in the development of vaccine strategies against bovine diseases caused by intracellular pathogens.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bromophenol Blue, titration: suitable