Merck
Hyperbaric oxygen promotes malignant glioma cell growth and inhibits cell apoptosis.
Oncology letters (2015-07-15)
Yong-Gang Wang, Yi-Ping Zhan, Shu-Yi Pan, Hai-Dong Wang, Dun-Xiao Zhang, Kai Gao, Xue-Ling Qi, Chun-Jiang Yu
ABSTRAKT
Glioblastoma multiforme (GBM) is the most frequently diagnosed intracranial malignant tumor in adults. Clinical studies have indicated that hyperbaric oxygen may improve the prognosis and reduce complications in glioma patients; however, the specific mechanism by which this occurs remains unknown. The present study investigated the direct effects of hyperbaric oxygen stimulation on glioma by constructing an intracranial transplanted glioma model in congenic C57BL/6J mice. Bioluminescent imaging (BLI) was used to assess the growth of intracranial transplanted GL261-Luc glioma cells
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC