Merck
Developmental immunotoxicity testing of 4-methyl anisole.
Regulatory toxicology and pharmacology : RTP (2015-04-18)
Elisa C M Tonk, Aart Verhoef, Eric R Gremmer, Henk van Loveren, Aldert H Piersma
ABSTRAKT
The developmental immunotoxicity of 4-methyl anisole (4MA) was investigated in the rat. Four study designs were used, with either premating or post-weaning onset of exposure, continued to postnatal day 50, and with or without additional oral gavage of pups from postnatal day 10 onward. Reduced litter size (benchmark dose lower confidence limit (BMDL) 80mg/kg bw/day) was the most sensitive developmental parameter, with pup relative organ weight effects observed at similar BMDLs, in the absence of maternal toxicity. Eosinophil numbers were reduced at lower doses (BMDL 16mg/kg bw/day). KLH challenge resulted in increased IL-13 and TNF-α responses, and variably reduced IgG production (BMDL 27mg/kg bw/day). T4 levels were reduced by 11% at maximum with a BMDL of 73mg/kg bw/day. Differences between exposure cohorts were limited and were considered to be without biological significance. This study shows that 4MA induces developmental immunotoxicity at doses below those inducing developmental and general toxicity. These observations being independent of the study designs applied suggest that the post-weaning period, included in all designs, is the most relevant sensitive period for inducing 4MA mediated developmental immunotoxicity. Moreover, this study stresses the importance of including developmental immunotoxicity testing by default in regulatory toxicology.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
4-Methylanisole, 99%
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis