Merck
  • Flotillin-2 promotes nasopharyngeal carcinoma metastasis and is necessary for the epithelial-mesenchymal transition induced by transforming growth factor-β.
Flotillin-2 promotes nasopharyngeal carcinoma metastasis and is necessary for the epithelial-mesenchymal transition induced by transforming growth factor-β.
Oncotarget (2015-04-25)
Liang Zhao, Li Lin, Changqie Pan, Min Shi, Yulin Liao, Jianping Bin, Wangjun Liao
ABSTRAKT
Transforming growth factor-β (TGF-β) promotes cancer metastasis via the epithelial-mesenchymal transition (EMT) but the underlying mechanisms in nasopharyngeal carcinoma (NPC) remain unclear. Flotillin-2 (Flot2), a specialized lipid raft domain in cellular membrane, was reported to promote cancer metastasis. Recently, in neuropathy, it was also suggested that Flot2 was involved in Src activation, which is known as the downstream signal of TGF-β. Therefore, we intended to find out the relationship between Flot2 and TGF-β in the process of nasopharyngeal carcinoma (NPC) metastasis. In this study, we found that Flot2 expression level positively correlated with the cancer stage in NPC tissues. Elevated Flot2 in tumor tissue was an independent prognostic marker, and higher Flot2 expression level showed shorter overall survival time in 181 NPC patients. In NPC cells, silencing Flot2 reversed the metastatic effect induced by TGF-β. Moreover, TGF-β-induced Src phosphorylation was significantly inhibited by Flot2 knocking down. As the consequence of Flot2 inhibition, the expression of the epithelial biomarker E-cadherin was upregulated, while the mesenchymal marker vimentin and signaling transducer β-catenin was suppressed. In conclusions, Flot2 is an indispensable member for TGF-β signaling, which is essential for the EMT process in NPC metastasis. Suppressing Flot2 may be a novel way against TGF-β-induced EMT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source