Merck
Effect of shear forces and ageing on the compliance of adhesive pads in adult cockroaches.
The Journal of experimental biology (2015-07-25)
Yanmin Zhou, Adam Robinson, Christine Viney, Walter Federle
ABSTRAKT
The flexibility of insect adhesive pads is crucial for their ability to attach on rough surfaces. Here, we used transparent substrates with micropillars to test in adult cockroaches (Nauphoeta cinerea) whether and how the stiffness of smooth adhesive pads changes when shear forces are applied, and whether the insect's age has any influence. We found that during pulls towards the body, the pad's ability to conform to the surface microstructures was improved in comparison to a contact without shear, suggesting that shear forces make the pad more compliant. The mechanism underlying this shear-dependent increase in compliance is still unclear. The effect was not explained by viscoelastic creep, changes in normal pressure, or shear-induced pad rolling, which brings new areas of cuticle into surface contact. Adhesive pads were significantly stiffer in older cockroaches. Stiffness increased most rapidly in cockroaches aged between 2.5 and 4 months. This increase is probably based on wear and repair of the delicate adhesive cuticle. Recent wear (visualised by Methylene Blue staining) was not age dependent, whereas permanent damage (visible as brown scars) accumulated with age, reducing the pads' flexibility.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution