Merck
Determination of seven selected neuro- and immunomodulatory steroids in human cerebrospinal fluid and plasma using LC-MS/MS.
Steroids (2015-02-14)
Lucie Sosvorova, Jana Vitku, Tereza Chlupacova, Milan Mohapl, Richard Hampl
ABSTRAKT
Dehydroepiandrosterone (DHEA) and its 7-oxo- and 7-hydroxy-metabolites occurring in the brain are considered neurosteroids. Metabolism of the latter is catalysed by 11β-hydroxysteroid dehydrogenase (11β-HSD) which also interconverts cortisol and cortisone. The concurrent metabolic reaction to DHEA 7-hydroxylation is the formation of 16α-hydroxy-DHEA. The LC-MS/MS method using triple stage quadrupole-mass spectrometer was developed for simultaneous quantification of free DHEA, 7α-hydroxy-DHEA, 7β-hydroxy-DHEA, 7-oxo-DHEA, 16α-hydroxy-DHEA, cortisol and cortisone in human plasma and cerebrospinal fluid (CSF). The method employs 500 μL of human plasma and 3000 μL of CSF extracted with diethyl ether and derivatized with 2-hydrazinopyridine. It has been validated in terms of sensitivity, precision and recovery. In plasma, the following values were obtained: limit of detection: 2-50p g/mL; limit of quantification: 5-140 pg/mL; within-day precision 0.58-14.58%; between-day precision: 1.24-13.89% and recovery: 85-113.2%). For CSF, the values of limit of detection: 2-28 pg/mL; limit of quantification: 6-94 pg/mL; within-day precision; 0.63-5.48%; between-day precision: 0.88-14.59% and recovery: 85.1-109.4% were acquired. Medians and concentration ranges of detected steroids in plasma and CSF are given in subjects with excluded normal pressure hydrocephalus (n=37; 65-80 years). The method enables simultaneous quantification of steroids important for the estimation of 11β-HSD activity in human plasma and CSF. It will be helpful in better understanding various degenerative diseases development and progression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Cortisone, ≥95%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)