Merck
  • A cost-effective and efficient reprogramming platform for large-scale production of integration-free human induced pluripotent stem cells in chemically defined culture.
A cost-effective and efficient reprogramming platform for large-scale production of integration-free human induced pluripotent stem cells in chemically defined culture.
Scientific reports (2015-06-13)
Jeanette Beers, Kaari L Linask, Jane A Chen, Lauren I Siniscalchi, Yongshun Lin, Wei Zheng, Mahendra Rao, Guokai Chen
ABSTRAKT
Factors limiting the adoption of iPSC technology include the cost of developing lines and the time period that it takes to characterize and bank them, particularly when integration free, feeder free, and Xeno-free components are used. In this manuscript we describe our optimization procedure that enables a single technician to make 20-40 lines at a time in a 24-96 well format in a reliable and reproducible fashion. Improvements spanned the entire workflow and included using RNA virus, reducing cytotoxicity of reagents, developing improved transfection and freezing efficiencies, modifying the manual colony picking steps, enhancing passaging efficiency and developing early criteria of success. These modifications allowed us to make more than two hundred well-characterized lines per year.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
Milli-Mark® Przeciwciało anty-Nanog-Alexa Fluor 488, NT, Milli-Mark®, from rabbit
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, FITC conjugate, clone TRA-1-60, from mouse, FITC conjugate
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, BioUltra, ≥98.0% (RT)
Sigma-Aldrich
Kontrola ujemna IgG2b myszy, klon GC198, koniugat FITC, Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.