Merck
Generation and tumor recognition properties of two human monoclonal antibodies specific to cell surface anionic phospholipids.
Investigational new drugs (2015-05-20)
Emil Bujak, Francesca Pretto, Dario Neri
ABSTRAKT
Phosphatidylserine (PS) and other anionic phospholipids, which become exposed on the surface of proliferating endothelial cells, tumor cells and certain leukocytes, have been used as targets for the development of clinical-stage biopharmaceuticals. One of these products (bavituximab) is currently being investigated in Phase 3 clinical trials. There are conflicting reports on the ability of bavituximab and other antibodies to recognize PS directly or through beta-2 glycoprotein 1, a serum protein that is not highly conserved across species. Here, we report on the generation and characterization of two fully human antibodies directed against phosphatidylserine. One of these antibodies (PS72) bound specifically to phosphatidylserine and to phosphatidic acid, but did not recognize other closely related phospholipids, while the other antibody (PS41) also bound to cardiolipin. Both PS72 and PS41 stained 8/9 experimental tumor models in vitro, but both antibodies failed to exhibit a preferential tumor accumulation in vivo, as revealed by quantitative biodistribution analysis. Our findings indicate that anionic phospholipids are exposed and accessible in most tumor types, but cast doubts about the possibility of efficiently targeting tumors in vivo with PS-specific reagents.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Carbon, mesoporous, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Carbon, mesoporous, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Węgiel - Szklisty, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Węgiel - Szklisty, foil, 10x10mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, rod, 50mm, diameter 1.0mm, glassy carbon
Węgiel - Szklisty, foil, 25x25mm, thickness 0.5mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, rod, 200mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, foil, 25x25mm, thickness 4.0mm, glassy carbon
Węgiel - Szklisty, rod, 5mm, diameter 3.0mm, glassy carbon
Węgiel - Szklisty, rod, 100mm, diameter 5.0mm, glassy carbon
Węgiel - Szklisty, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Węgiel - Szklisty, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.99 atom % 12C
Sigma-Aldrich
DAPI, for nucleic acid staining