Merck
Raman Characterization of the UV-Protective Pigment Gloeocapsin and Its Role in the Survival of Cyanobacteria.
Astrobiology (2015-09-26)
Jean-Yves Storme, Stjepko Golubic, Annick Wilmotte, Julia Kleinteich, David Velázquez, Emmanuelle J Javaux
ABSTRAKT
Extracellular UV-screening pigments gloeocapsin and scytonemin present in the exopolysaccharide (EPS) envelopes of extremophilic cyanobacteria of freshwater and marine environments were studied by Raman spectroscopy and compared to their intracellular photosynthetic pigments. This Raman spectral analysis of the extracellular pigment gloeocapsin showed that it shared Raman spectral signatures with parietin, a radiation-protective pigment known in lichens. The UV-light spectra also show similarities. Gloeocapsin occurs in some cyanobacterial species, mostly with exclusion of scytonemin, indicating that these pigments have evolved in cyanobacteria as separate protective strategies. Both gloeocapsin and scytonemin are widely and species-specifically distributed in different cyanobacterial genera and families. The widespread occurrence of these pigments may suggest an early origin, while their detection by Raman spectroscopy makes them potential biosignatures for cyanobacteria in the fossil record and demonstrates the usefulness of nondestructive Raman spectroscopy analyses for the search for complex organics, including possible photosynthetic pigments, if preservable in early Earth and extraterrestrial samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether