Merck
Clinical Characteristics and Bacteriological Profile of Moraxella Keratitis.
Cornea (2015-05-15)
Hidenori Inoue, Takashi Suzuki, Tomoyuki Inoue, Takaaki Hattori, Ryohei Nejima, Daisuke Todokoro, Saichi Hoshi, Hiroshi Eguchi, Hitoshi Miyamoto, Yuichi Ohashi
ABSTRAKT
Moraxella species are rare causative pathogens of severe sight-threatening keratitis. The aim of this study was to analyze the clinical presentation, predisposing risk factors, in vitro antimicrobial susceptibility, and treatment associated with Moraxella keratitis. We retrospectively reviewed 30 culture-proven cases of Moraxella keratitis from multiple centers in Japan. The mean age of the patients was 58.4 ± 23.4 years. The most common ocular conditions were contact lens wearing (5 patients, 16.7%) and trauma (3 patients, 10.0%). Seven patients had diabetes mellitus. Sixteen patients exhibited hypopyon in association with the corneal focus. Ring-shaped infiltration was found in 9 patients (30.0%), and irregular or amoebic-shaped infiltration was observed in 13 patients (43.3%). Eight patients (26.7%) showed small round infiltrates. All Moraxella isolates were sensitive to fluoroquinolones and aminoglycosides. All were treated with a combination ophthalmic solution containing a fluoroquinolone, tobramycin, and cefmenoxime. Although no patients developed corneal perforation, the response to treatment was slow in all cases; the mean treatment period was 41.9 days. In Japan, Moraxella keratitis occurs in patients with contact lens wear, trauma, and diabetes mellitus. It presents as a small, round, ring-shaped, irregularly shaped, or amoebic-shaped focus. Moraxella species exhibit good susceptibility to fluoroquinolones and aminoglycosides. Because the treatment response may be very slow, these agents should be continued for a long period of time.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%